Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PDE11A
Entrez Gene ID
50940
Gene Name
phosphodiesterase 11A
Gene Aliases
PPNAD2
Chromosome Location
Chr.2: 177623249 - 178108339 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001077196.1 NP_001070664.1 8 - 9 757 128
NM_001077197.1 NP_001070665.1 12 - 13 1287 128
NM_001077358.1 NP_001070826.1 10 - 11 997 128
NM_016953.3 NP_058649.3 11 - 12 2256 128
GenBank mRNA AB036704.1 - 11 - 12 2256 128
AB038041.1 - 12 - 13 1287 128
AF281865.1 - 10 - 11 969 128
AJ251509.1 - 8 - 9 756 128
AJ278682.1 - 12 - 13 1330 128
AK074273.1 - 1 - 2 153 128
BC112393.1 - 11 - 12 2247 128
BC114431.1 - 11 - 12 2247 128
BX649056.1 - 10 - 11 981 128
HM771389.1 - 11 - 12 1938 128
HM771390.1 - 11 - 12 1938 128
HM771391.1 - 11 - 12 1938 128

View Full Product Details