The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001077196.1 | NP_001070664.1 | 9 - 10 | 865 | 67 |
NM_001077197.1 | NP_001070665.1 | 13 - 14 | 1395 | 67 | |
NM_001077358.1 | NP_001070826.1 | 11 - 12 | 1105 | 67 | |
NM_016953.3 | NP_058649.3 | 12 - 13 | 2364 | 67 | |
GenBank mRNA | AB036704.1 | - | 12 - 13 | 2364 | 67 |
AB038041.1 | - | 13 - 14 | 1395 | 67 | |
AF281865.1 | - | 11 - 12 | 1077 | 67 | |
AJ251509.1 | - | 9 - 10 | 864 | 67 | |
AJ278682.1 | - | 13 - 14 | 1438 | 67 | |
AK074273.1 | - | 2 - 3 | 261 | 67 | |
BC112393.1 | - | 12 - 13 | 2355 | 67 | |
BC114431.1 | - | 12 - 13 | 2355 | 67 | |
BX649056.1 | - | 11 - 12 | 1089 | 67 | |
HM771389.1 | - | 12 - 13 | 2046 | 67 | |
HM771390.1 | - | 12 - 13 | 2046 | 67 | |
HM771391.1 | - | 12 - 13 | 2046 | 67 |