Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PDE11A
Entrez Gene ID
50940
Gene Name
phosphodiesterase 11A
Gene Aliases
PPNAD2
Chromosome Location
Chr.2: 177623249 - 178108339 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001077196.1 NP_001070664.1 13 - 14 1242 127
NM_001077197.1 NP_001070665.1 17 - 18 1772 127
NM_001077358.1 NP_001070826.1 15 - 16 1482 127
NM_016953.3 NP_058649.3 16 - 17 2741 127
GenBank mRNA AB036704.1 - 16 - 17 2741 127
AB038041.1 - 17 - 18 1772 127
AF281865.1 - 15 - 16 1454 127
AJ251509.1 - 13 - 14 1241 127
AJ278682.1 - 17 - 18 1815 127
AK074273.1 - 6 - 7 639 128
BC112393.1 - 16 - 17 2732 127
BC114431.1 - 16 - 17 2732 127
HM771389.1 - 16 - 17 2423 127
HM771390.1 - 16 - 17 2423 127
HM771391.1 - 16 - 17 2423 127

View Full Product Details