Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SCN1A
Entrez Gene ID
6323
Gene Name
sodium voltage-gated channel alpha subunit 1
Gene Aliases
EIEE6, FEB3, FEB3A, FHM3, GEFSP2, HBSCI, NAC1, Nav1.1, SCN1, SMEI
Chromosome Location
Chr.2: 165989160 - 166149216 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_006920.4 NP_008851.3 11 - 12 2031 95
XM_011511604.2 XP_011509906.1 12 - 13 2200 95
XM_011511605.2 XP_011509907.1 13 - 14 2404 95
XM_017004645.1 XP_016860134.1 13 - 14 2407 95
XM_017004646.1 XP_016860135.1 12 - 13 2321 95
XM_017004647.1 XP_016860136.1 14 - 15 2493 95
XM_017004648.1 XP_016860137.1 13 - 14 2270 95
XM_017004649.1 XP_016860138.1 12 - 13 2232 95
XM_017004650.1 XP_016860139.1 12 - 13 2306 95
XM_017004654.1 XP_016860143.1 13 - 14 2423 95
GenBank mRNA AB093549.1 - 11 - 12 2013 95
AY043484.1 - 11 - 12 2013 95

View Full Product Details