The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_138477.2 | NP_612486.2 | 20 - 21 | 2928 | 97 |
XM_005254176.4 | XP_005254233.1 | 20 - 21 | 2843 | 97 | |
XM_011521270.2 | XP_011519572.1 | 20 - 21 | 2868 | 97 | |
XM_011521271.2 | XP_011519573.1 | 20 - 21 | 2865 | 97 | |
XM_011521274.2 | XP_011519576.1 | 17 - 18 | 1914 | 97 | |
GenBank mRNA | AF525398.1 | - | 20 - 21 | 2925 | 97 |
BC001092.2 | - | 12 - 13 | 1383 | 97 | |
BC008333.1 | - | 12 - 13 | 1383 | 97 | |
BC008334.1 | - | 12 - 13 | 1383 | 97 | |
BC052568.1 | - | 20 - 21 | 2816 | 97 | |
BC066640.1 | - | 20 - 21 | 2816 | 97 |