Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TBC1D10A
Entrez Gene ID
83874
Gene Name
TBC1 domain family member 10A
Gene Aliases
EPI64, TBC1D10, dJ130H16.1, dJ130H16.2
Chromosome Location
Chr.22: 30291990 - 30326966 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031937.2 NP_114143.1 1 - 2 294 110
GenBank mRNA AB449894.1 - 1 - 2 209 110
AF331038.1 - 1 - 2 233 110
AK074656.1 - 1 - 2 260 110
BC022201.1 - 1 - 2 243 110
BC136815.1 - 1 - 2 250 110
BC142940.1 - 1 - 2 266 110
BC150187.1 - 1 - 2 214 110
BC150214.1 - 1 - 2 219 110
BC171775.1 - 1 - 2 250 110
CT841514.1 - 1 - 2 215 110
DA944522.1 - 1 - 2 297 110

View Full Product Details