The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001252338.2 | NP_001239267.1 | 8 - 9 | 1282 | 92 |
NM_001252339.2 | NP_001239268.1 | 8 - 9 | 1439 | 92 | |
NM_001794.4 | NP_001785.2 | 9 - 10 | 1467 | 92 | |
GenBank mRNA | AK091496.1 | - | 9 - 10 | 1663 | 92 |
AK124724.1 | - | 8 - 9 | 1422 | 92 | |
AK308345.1 | - | 8 - 9 | 1281 | 92 | |
AK315192.1 | - | 9 - 10 | 1421 | 92 | |
BC101651.1 | - | 9 - 10 | 1467 | 92 | |
BC112150.1 | - | 9 - 10 | 1467 | 92 | |
L34059.1 | - | 9 - 10 | 1393 | 92 |