Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HGF
Entrez Gene ID
3082
Gene Name
hepatocyte growth factor
Gene Aliases
DFNB39, F-TCF, HGFB, HPTA, SF
Chromosome Location
Chr.7: 81699006 - 81770438 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000601.5 NP_000592.3 16 - 17 2092 112
NM_001010932.2 NP_001010932.1 16 - 17 2077 112
XM_006715956.2 XP_006716019.1 17 - 18 2055 112
XM_011516115.2 XP_011514417.1 17 - 18 2040 112
GenBank mRNA AK291674.1 - 16 - 17 2027 112
AK313694.1 - 16 - 17 1927 112
BC130284.1 - 16 - 17 1940 112
BC130286.1 - 16 - 17 1940 112
M29145.1 - 16 - 17 1966 112
M55379.1 - 16 - 17 1850 112
M60718.1 - 16 - 17 1937 112
M73239.1 - 16 - 17 1930 112
M73240.1 - 16 - 17 1915 112
X16323.1 - 16 - 17 1999 112

View Full Product Details