Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BRF1
Entrez Gene ID
2972
Gene Name
BRF1, RNA polymerase III transcription initiation factor 90 kDa subunit
Gene Aliases
BRF, BRF-1, CFDS, GTF3B, HEL-S-76p, TAF3B2, TAF3C, TAFIII90, TF3B90, TFIIIB90, hBRF
Chromosome Location
Chr.14: 105209286 - 105315577 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001242786.1 NP_001229715.1 17 - 18 2132 59
NM_001242787.1 NP_001229716.1 17 - 18 2066 59
NM_001242788.1 NP_001229717.1 16 - 17 2276 59
NM_001242789.1 NP_001229718.1 12 - 13 1556 59
NM_001519.3 NP_001510.2 17 - 18 2357 59
NM_145685.2 NP_663718.1 13 - 14 2265 59
XM_005267561.3 XP_005267618.1 17 - 18 2376 59
XM_005267563.3 XP_005267620.1 13 - 14 3069 59
XM_006720123.2 XP_006720186.1 12 - 13 2497 59
XM_011536673.1 XP_011534975.1 16 - 17 2229 59
XM_011536674.1 XP_011534976.1 13 - 14 2647 59
GenBank mRNA AJ297407.1 - 13 - 14 2273 59
AK096471.1 - 12 - 13 1556 59
AK124665.1 - 7 - 8 1421 59
AK125024.1 - 4 - 5 1871 59
AK294899.1 - 17 - 18 2132 59
AK295579.1 - 17 - 18 1979 59
BC071637.1 - 12 - 13 1421 59
BC086856.1 - 16 - 17 2009 59
U28838.1 - 17 - 18 2354 59
U75276.1 - 17 - 18 2061 59

View Full Product Details