The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001271004.1 | NP_001257933.1 | 1 - 2 | 406 | 81 |
NM_006528.3 | NP_006519.1 | 1 - 2 | 406 | 81 | |
GenBank mRNA | AK024817.1 | - | 1 - 2 | 168 | 81 |
AK075425.1 | - | 1 - 2 | 168 | 81 | |
AK075427.1 | - | 1 - 2 | 167 | 81 | |
AK129833.1 | - | 1 - 2 | 168 | 81 | |
AK313260.1 | - | 1 - 2 | 168 | 81 | |
AY691946.1 | - | 1 - 2 | 144 | 81 | |
BC005330.1 | - | 1 - 2 | 168 | 81 | |
D29992.1 | - | 1 - 2 | 149 | 81 | |
DA831865.1 | - | 1 - 2 | 406 | 81 | |
DA833200.1 | - | 1 - 2 | 167 | 81 | |
L27624.1 | - | 1 - 2 | 131 | 81 |