Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PLEKHG1
Entrez Gene ID
57480
Gene Name
pleckstrin homology and RhoGEF domain containing G1
Gene Aliases
ARHGEF41
Chromosome Location
Chr.6: 150599863 - 150843665 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001029884.1 NP_001025055.1 14 - 15 1765 106
XM_005267064.3 XP_005267121.1 13 - 14 3844 106
XM_005267065.3 XP_005267122.1 15 - 16 1791 106
XM_005267066.2 XP_005267123.1 13 - 14 2648 106
XM_006715521.1 XP_006715584.1 15 - 16 2018 106
XM_006715522.3 XP_006715585.1 13 - 14 3885 106
XM_011535981.2 XP_011534283.1 13 - 14 4641 106
XM_011535983.2 XP_011534285.1 14 - 15 3066 106
GenBank mRNA AB033035.1 - 7 - 8 638 106
AJ420468.1 - 11 - 12 1003 106
AK056300.1 - 13 - 14 1664 106
AK293288.1 - 13 - 14 1696 106
AK294798.1 - 13 - 14 1819 106
AK307631.1 - 13 - 14 1718 106
BC089428.1 - 9 - 10 876 106
BC140864.1 - 12 - 13 1502 106
BC151134.1 - 12 - 13 1502 106

View Full Product Details