Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ACTR10
Entrez Gene ID
55860
Gene Name
actin-related protein 10 homolog
Gene Aliases
ACTR11, Arp10, Arp11, HARP11
Chromosome Location
Chr.14: 58200115 - 58235636 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018477.2 NP_060947.1 10 - 11 889 152
XM_011536960.1 XP_011535262.1 10 - 11 855 152
XM_011536961.1 XP_011535263.1 9 - 10 798 152
XM_011536962.2 XP_011535264.1 8 - 9 691 152
XM_011536963.2 XP_011535265.1 8 - 9 669 152
GenBank mRNA AF220190.1 - 10 - 11 866 152
AK000544.1 - 10 - 11 869 152
AK022248.1 - - 1083 152
AK022534.1 - 10 - 11 841 152
AK222624.1 - 10 - 11 850 152
AK222702.1 - 10 - 11 850 152
AK315537.1 - 10 - 11 834 152
BC005208.1 - 10 - 11 850 152
BC011997.1 - 10 - 11 823 152
BX161491.1 - 5 - 6 933 152

View Full Product Details