The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018477.2 | NP_060947.1 | 4 - 5 | 443 | 125 |
XM_011536960.1 | XP_011535262.1 | 4 - 5 | 388 | 125 | |
XM_011536961.1 | XP_011535263.1 | 4 - 5 | 388 | 125 | |
GenBank mRNA | AF220190.1 | - | 4 - 5 | 420 | 125 |
AK022248.1 | - | - | 673 | 125 | |
AK022534.1 | - | 4 - 5 | 395 | 125 | |
AK222624.1 | - | 4 - 5 | 404 | 125 | |
AK222702.1 | - | 4 - 5 | 404 | 125 | |
AK315537.1 | - | 4 - 5 | 388 | 125 | |
BC005208.1 | - | 4 - 5 | 404 | 125 | |
BC011997.1 | - | 4 - 5 | 377 | 125 | |
BX248282.1 | - | 4 - 5 | 339 | 125 |