Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ACTR10
Entrez Gene ID
55860
Gene Name
actin-related protein 10 homolog
Gene Aliases
ACTR11, Arp10, Arp11, HARP11
Chromosome Location
Chr.14: 58200115 - 58235636 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018477.2 NP_060947.1 5 - 6 551 86
XM_011536960.1 XP_011535262.1 5 - 6 496 86
XM_011536961.1 XP_011535263.1 5 - 6 496 86
GenBank mRNA AF220190.1 - 5 - 6 528 86
AK000544.1 - 5 - 6 531 86
AK022248.1 - - 781 86
AK022534.1 - 5 - 6 503 86
AK222624.1 - 5 - 6 512 86
AK222702.1 - 5 - 6 512 86
AK315537.1 - 5 - 6 496 86
BC005208.1 - 5 - 6 512 86
BC011997.1 - 5 - 6 485 86
BX248282.1 - 5 - 6 447 86

View Full Product Details