The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001001521.1 | NP_001001521.1 | 5 - 6 | 826 | 107 |
NM_006759.3 | NP_006750.3 | 5 - 6 | 882 | 107 | |
XM_005264537.2 | XP_005264594.1 | 6 - 7 | 934 | 107 | |
XM_005264538.1 | XP_005264595.1 | 6 - 7 | 901 | 107 | |
XM_006712088.1 | XP_006712151.1 | 6 - 7 | 1007 | 107 | |
XM_011533091.1 | XP_011531393.1 | 3 - 4 | 561 | 107 | |
XM_017004855.1 | XP_016860344.1 | 6 - 7 | 902 | 107 | |
XM_017004856.1 | XP_016860345.1 | 7 - 8 | 794 | 107 | |
XM_017004857.1 | XP_016860346.1 | 7 - 8 | 1095 | 107 | |
GenBank mRNA | AK300731.1 | - | 5 - 6 | 610 | 107 |
AK314263.1 | - | 5 - 6 | 873 | 107 | |
BC000173.2 | - | 5 - 6 | 827 | 107 | |
BC002954.1 | - | 5 - 6 | 706 | 107 | |
BC047004.1 | - | 5 - 6 | 833 | 107 | |
BX647825.1 | - | 3 - 4 | 674 | 107 | |
U27460.1 | - | 6 - 7 | 651 | 107 |