Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- CELF4
- Entrez Gene ID
-
56853
- Gene Name
- CUGBP, Elav-like family member 4
- Gene Aliases
-
BRUNOL4, CELF-4
- Chromosome Location
-
Chr.18: 37243040 - 37566052 on Build GRCh38
View Full Product Details