The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_023034.1 | NP_075447.1 | 10 - 11 | 2508 | 65 |
GenBank mRNA | AF332469.1 | - | 10 - 11 | 2508 | 65 |
AJ295990.1 | - | 10 - 11 | 2303 | 65 | |
AJ295991.1 | - | 10 - 11 | 2303 | 65 | |
BC101717.1 | - | 10 - 11 | 2082 | 65 | |
BC113469.1 | - | 10 - 11 | 2082 | 65 | |
BC115006.1 | - | 10 - 11 | 2103 | 65 | |
BC143510.1 | - | 10 - 11 | 2082 | 65 |