The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032351.5 | NP_115727.5 | 6 - 7 | 819 | 58 |
GenBank mRNA | AK172741.1 | - | 6 - 7 | 679 | 58 |
AK225031.1 | - | 6 - 7 | 679 | 58 | |
AK293605.1 | - | 6 - 7 | 785 | 58 | |
BC130382.1 | - | 6 - 7 | 710 | 58 | |
BC130384.1 | - | 6 - 7 | 710 | 58 |