Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC36A4
Entrez Gene ID
120103
Gene Name
solute carrier family 36 member 4
Gene Aliases
PAT4
Chromosome Location
Chr.11: 93144171 - 93197983 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_152313.3 NP_689526.2 3 - 4 413 115
XM_017017174.1 XP_016872663.1 3 - 4 417 115
GenBank mRNA AY162216.1 - 3 - 4 367 115
BC047374.1 - 3 - 4 380 115
HY170772.1 - 3 - 4 413 115

View Full Product Details