The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_182488.3 | NP_872294.2 | 7 - 8 | 1194 | 103 |
XM_011534956.2 | XP_011533258.1 | 7 - 8 | 945 | 103 | |
XM_017020416.1 | XP_016875905.1 | 6 - 7 | 865 | 103 | |
GenBank mRNA | AF022789.1 | - | 7 - 8 | 1044 | 103 |
AK289685.1 | - | 7 - 8 | 1108 | 103 | |
BC026072.1 | - | 7 - 8 | 1195 | 103 |