Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TEX30
Entrez Gene ID
93081
Gene Name
testis expressed 30
Gene Aliases
C13orf27
Chromosome Location
Chr.13: 102765888 - 102773821 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001286776.1 NP_001273705.1 3 - 4 425 69
NM_138779.4 NP_620134.3 4 - 5 500 69
XM_005254097.2 XP_005254154.1 4 - 5 554 69
XM_011521135.2 XP_011519437.1 4 - 5 1141 69
XM_011521136.2 XP_011519438.1 2 - 3 270 69
XM_017020852.1 XP_016876341.1 4 - 5 1485 69
GenBank mRNA AF070559.1 - 2 - 3 1014 69
AK314995.1 - 4 - 5 429 69
BC015148.2 - 4 - 5 471 69
BC048011.1 - 1 - 2 5532 69
BI559423.1 - 3 - 4 398 69
BP363423.1 - 3 - 4 403 69
HY045638.1 - 3 - 4 425 69

View Full Product Details