The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012291.4 | NP_036423.4 | 15 - 16 | 3016 | 71 |
XM_006719705.3 | XP_006719768.1 | 15 - 16 | 3083 | 71 | |
XM_011539024.2 | XP_011537326.1 | 15 - 16 | 3044 | 71 | |
XM_011539025.2 | XP_011537327.1 | 15 - 16 | 3055 | 71 | |
XM_017020253.1 | XP_016875742.1 | 15 - 16 | 3134 | 71 | |
GenBank mRNA | AK299423.1 | - | 4 - 5 | 588 | 71 |
AK316135.1 | - | 4 - 5 | 588 | 71 | |
AY455930.1 | - | 14 - 15 | 2925 | 71 | |
BC047603.1 | - | 13 - 14 | 1814 | 71 | |
D79987.1 | - | 15 - 16 | 3063 | 71 |