The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001323573.1 | NP_001310502.1 | 10 - 11 | 1305 | 60 |
NM_001323574.1 | NP_001310503.1 | 10 - 11 | 1305 | 60 | |
NM_001323575.1 | NP_001310504.1 | 10 - 11 | 1269 | 60 | |
NM_032785.3 | NP_116174.3 | 10 - 11 | 1269 | 60 | |
NR_136623.1 | 10 - 11 | 1268 | 60 | ||
XM_011542308.2 | XP_011540610.1 | 11 - 12 | 1364 | 60 | |
XM_017002595.1 | XP_016858084.1 | 10 - 11 | 1326 | 60 | |
XM_017002596.1 | XP_016858085.1 | 10 - 11 | 1326 | 60 | |
XM_017002597.1 | XP_016858086.1 | 10 - 11 | 1326 | 60 | |
GenBank mRNA | AK094826.1 | - | 10 - 11 | 1228 | 60 |
AK294394.1 | - | 10 - 11 | 1264 | 60 | |
AK294633.1 | - | 10 - 11 | 1268 | 60 | |
AK296221.1 | - | 9 - 10 | 1085 | 60 | |
BC126383.1 | - | 6 - 7 | 581 | 60 | |
BC144171.1 | - | 5 - 6 | 491 | 60 |