Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
OSBPL9
Entrez Gene ID
114883
Gene Name
oxysterol binding protein like 9
Gene Aliases
ORP-9, ORP9
Chromosome Location
Chr.1: 51616874 - 51789219 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024586.5 NP_078862.4 17 - 18 1747 125
NM_148904.3 NP_683702.1 13 - 14 1464 125
NM_148905.3 NP_683703.1 12 - 13 1368 125
NM_148906.2 NP_683704.2 16 - 17 1696 125
NM_148907.2 NP_683705.1 13 - 14 1341 125
NM_148908.3 NP_683706.3 16 - 17 1708 125
NM_148909.3 NP_683707.3 17 - 18 1777 125
NR_036662.1 16 - 17 1668 125
XM_006710318.2 XP_006710381.1 16 - 17 1507 125
XM_006710320.3 XP_006710383.1 14 - 15 1344 125
XM_006710321.2 XP_006710384.1 13 - 14 1340 125
XM_006710322.3 XP_006710385.1 14 - 15 1501 125
XM_006710323.2 XP_006710386.1 13 - 14 1409 125
XM_006710324.3 XP_006710387.1 15 - 16 1419 125
XM_006710325.3 XP_006710388.1 14 - 15 1389 125
XM_006710326.2 XP_006710389.1 12 - 13 1391 125
XM_011540599.2 XP_011538901.1 18 - 19 1601 125
XM_011540600.2 XP_011538902.1 17 - 18 1568 125
XM_011540601.2 XP_011538903.1 16 - 17 1529 125
XM_011540602.2 XP_011538904.1 17 - 18 1503 125
XM_011540603.2 XP_011538905.1 17 - 18 1527 125
XM_011540604.2 XP_011538906.1 14 - 15 1302 125
XM_017000218.1 XP_016855707.1 15 - 16 1468 125
XM_017000219.1 XP_016855708.1 16 - 17 1488 125
XM_017000220.1 XP_016855709.1 13 - 14 1229 125
XM_017000221.1 XP_016855710.1 13 - 14 1180 125
XM_017000222.1 XP_016855711.1 17 - 18 1713 125
XM_017000223.1 XP_016855712.1 17 - 18 1711 125
XM_017000224.1 XP_016855713.1 14 - 15 1656 125
XM_017000225.1 XP_016855714.1 15 - 16 1507 125
GenBank mRNA AB208851.1 - 5 - 6 928 125
AF392445.1 - 17 - 18 1521 125
AK022554.1 - 12 - 13 1148 125
AK027535.1 - 16 - 17 1478 125
AK027707.1 - 13 - 14 1304 125
AK056617.1 - 17 - 18 1566 125
AK074392.1 - 3 - 4 280 125
AK091703.1 - 16 - 17 1450 125
AK124037.1 - 8 - 9 1994 125
AK128043.1 - 19 - 20 1971 125
AK297656.1 - 14 - 15 1494 125
AK315167.1 - 13 - 14 1340 125
AY178997.1 - 16 - 17 1470 125
BC025978.2 - 13 - 14 1374 125
CR457288.1 - 10 - 11 1015 125

View Full Product Details