Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TUBGCP3
Entrez Gene ID
10426
Gene Name
tubulin gamma complex associated protein 3
Gene Aliases
104p, GCP3, Grip104, SPBC98, Spc98, Spc98p
Chromosome Location
Chr.13: 112485005 - 112606469 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001286278.1 NP_001273207.1 4 - 5 531 76
NM_001286279.1 NP_001273208.1 4 - 5 531 76
NM_006322.5 NP_006313.1 4 - 5 531 76
XM_005268293.1 XP_005268350.1 4 - 5 526 76
XM_011537462.2 XP_011535764.1 4 - 5 484 76
XM_011537463.2 XP_011535765.1 5 - 6 460 76
XM_011537464.2 XP_011535766.1 4 - 5 1264 76
XM_017020323.1 XP_016875812.1 5 - 6 878 76
GenBank mRNA AF042378.1 - 4 - 5 410 76
AJ003061.1 - 4 - 5 326 76
AJ003062.1 - 4 - 5 326 76
AK314785.1 - 4 - 5 531 76
BC007763.2 - 4 - 5 481 76
BC013781.1 - 4 - 5 471 76
BC046634.2 - 4 - 5 514 76
BT009772.1 - 4 - 5 326 76

View Full Product Details