The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001256796.1 | NP_001243725.1 | 8 - 9 | 1094 | 82 |
NM_001261397.1 | NP_001248326.1 | 7 - 8 | 965 | 82 | |
NM_001261398.1 | NP_001248327.1 | 7 - 8 | 1046 | 82 | |
NM_002560.2 | NP_002551.2 | 7 - 8 | 1046 | 82 | |
NR_046372.1 | 8 - 9 | 1084 | 82 | ||
NR_046373.1 | 6 - 7 | 898 | 82 | ||
XM_011538416.2 | XP_011536718.1 | 3 - 4 | 409 | 82 | |
GenBank mRNA | AF000234.1 | - | 7 - 8 | 642 | 82 |
AK225604.1 | - | 7 - 8 | 742 | 82 | |
AK293599.1 | - | 4 - 5 | 536 | 82 | |
AK293660.1 | - | 6 - 7 | 681 | 82 | |
AK297704.1 | - | 6 - 7 | 630 | 82 | |
AK301080.1 | - | 7 - 8 | 730 | 82 | |
AK308549.1 | - | 8 - 9 | 816 | 82 | |
AK308868.1 | - | 7 - 8 | 1737 | 82 | |
BC017458.1 | - | 7 - 8 | 775 | 82 | |
BC033826.1 | - | 7 - 8 | 766 | 82 | |
BT019738.1 | - | 7 - 8 | 742 | 82 | |
BT019739.1 | - | 7 - 8 | 661 | 82 | |
BX402887.2 | - | 8 - 9 | 864 | 82 | |
DQ851491.1 | - | 3 - 4 | 260 | 82 | |
U83993.1 | - | 7 - 8 | 1051 | 82 | |
Y07684.1 | - | 7 - 8 | 769 | 82 |