The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001256796.1 | NP_001243725.1 | 9 - 10 | 1234 | 73 |
NM_001261397.1 | NP_001248326.1 | 8 - 9 | 1105 | 73 | |
NM_001261398.1 | NP_001248327.1 | 8 - 9 | 1186 | 73 | |
NM_002560.2 | NP_002551.2 | 8 - 9 | 1186 | 73 | |
NR_046372.1 | 9 - 10 | 1224 | 73 | ||
NR_046373.1 | 7 - 8 | 1038 | 73 | ||
XM_011538416.2 | XP_011536718.1 | 4 - 5 | 549 | 73 | |
GenBank mRNA | AF000234.1 | - | 8 - 9 | 782 | 73 |
AF012903.1 | - | 7 - 8 | 668 | 73 | |
AK225604.1 | - | 8 - 9 | 882 | 73 | |
AK293599.1 | - | 5 - 6 | 676 | 73 | |
AK293660.1 | - | 7 - 8 | 821 | 73 | |
AK297704.1 | - | 7 - 8 | 770 | 73 | |
AK301080.1 | - | 8 - 9 | 870 | 73 | |
AK308549.1 | - | 9 - 10 | 956 | 73 | |
AK308868.1 | - | 8 - 9 | 1877 | 73 | |
BC017458.1 | - | 8 - 9 | 915 | 73 | |
BC033826.1 | - | 8 - 9 | 906 | 73 | |
BT019738.1 | - | 8 - 9 | 882 | 73 | |
BT019739.1 | - | 8 - 9 | 801 | 73 | |
JX471561.1 | - | 1 - 2 | 78 | 73 | |
U83993.1 | - | 8 - 9 | 1191 | 73 | |
Y07684.1 | - | 8 - 9 | 909 | 73 |