The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001314022.1 | NP_001300951.1 | 3 - 4 | 727 | 71 |
NM_001314023.1 | NP_001300952.1 | 4 - 5 | 399 | 71 | |
NM_001314024.1 | NP_001300953.1 | 3 - 4 | 270 | 71 | |
NM_001314025.1 | NP_001300954.1 | 4 - 5 | 534 | 71 | |
NM_058181.2 | NP_478061.1 | 4 - 5 | 856 | 71 | |
XM_005261156.3 | XP_005261213.2 | 3 - 4 | 679 | 71 | |
XM_017028395.1 | XP_016883884.1 | - | 2451 | 71 | |
GenBank mRNA | AI262212.1 | - | 1 - 2 | 71 | |
AK128875.1 | - | 4 - 5 | 468 | 71 | |
AY040873.1 | - | 4 - 5 | 802 | 71 | |
AY040874.1 | - | 3 - 4 | 673 | 71 | |
AY040875.1 | - | 4 - 5 | 345 | 71 | |
AY040876.1 | - | 3 - 4 | 216 | 71 | |
BG473705.1 | - | 3 - 4 | 440 | 71 |