Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WDR4
Entrez Gene ID
10785
Gene Name
WD repeat domain 4
Gene Aliases
TRM82, TRMT82
Chromosome Location
Chr.21: 42843075 - 42879865 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_033661.4 NP_387510.1 11 - 12 1333 63
XM_017028264.1 XP_016883753.1 9 - 10 990 63
GenBank mRNA AJ243912.1 - 12 - 13 1277 63
BC018866.2 - 3 - 4 429 63
CD368164.1 - 1 - 2 63

View Full Product Details