The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001260474.1 | NP_001247403.1 | 4 - 5 | 542 | 88 |
NM_001260475.1 | NP_001247404.1 | 4 - 5 | 490 | 88 | |
NM_001260476.1 | NP_001247405.1 | 4 - 5 | 390 | 88 | |
NM_001260477.1 | NP_001247406.1 | 4 - 5 | 597 | 88 | |
NM_018669.5 | NP_061139.2 | 4 - 5 | 542 | 88 | |
NM_033661.4 | NP_387510.1 | 4 - 5 | 542 | 88 | |
NR_048535.1 | 4 - 5 | 434 | 88 | ||
XM_011529433.1 | XP_011527735.1 | 4 - 5 | 805 | 88 | |
GenBank mRNA | AK056343.1 | - | 4 - 5 | 597 | 88 |
AK292923.1 | - | 4 - 5 | 483 | 88 | |
AK298015.1 | - | 4 - 5 | 434 | 88 | |
AK315008.1 | - | 4 - 5 | 433 | 88 | |
BC001074.2 | - | 4 - 5 | 369 | 88 | |
BC006341.2 | - | 4 - 5 | 490 | 88 | |
BU541429.1 | - | 4 - 5 | 468 | 88 | |
DC303088.1 | - | 4 - 5 | 490 | 88 |