The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024586.5 | NP_078862.4 | 3 - 4 | 482 | 82 |
NM_148906.2 | NP_683704.2 | 2 - 3 | 431 | 82 | |
NM_148908.3 | NP_683706.3 | 3 - 4 | 482 | 82 | |
NM_148909.3 | NP_683707.3 | 3 - 4 | 482 | 82 | |
XM_006710318.2 | XP_006710381.1 | 3 - 4 | 272 | 82 | |
XM_011540599.2 | XP_011538901.1 | 3 - 4 | 267 | 82 | |
XM_011540600.2 | XP_011538902.1 | 3 - 4 | 264 | 82 | |
XM_011540601.2 | XP_011538903.1 | 3 - 4 | 264 | 82 | |
XM_011540602.2 | XP_011538904.1 | 3 - 4 | 238 | 82 | |
XM_017000218.1 | XP_016855707.1 | 3 - 4 | 272 | 82 | |
XM_017000222.1 | XP_016855711.1 | 3 - 4 | 264 | 82 | |
GenBank mRNA | AF392445.1 | - | 3 - 4 | 256 | 82 |
AK027535.1 | - | 3 - 4 | 252 | 82 | |
AK056617.1 | - | 3 - 4 | 271 | 82 | |
AK128043.1 | - | 6 - 7 | 745 | 82 | |
AY178997.1 | - | 2 - 3 | 205 | 82 |