Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TNFSF13B
Entrez Gene ID
10673
Gene Name
tumor necrosis factor superfamily member 13b
Gene Aliases
BAFF, BLYS, CD257, DTL, TALL-1, TALL1, THANK, TNFSF20, TNLG7A, ZTNF4
Chromosome Location
Chr.13: 108268240 - 108308484 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_006573.4 NP_006564.1 2 - 3 687 81
GenBank mRNA AF116456.1 - 2 - 3 538 81
AF132600.1 - 2 - 3 556 81
AF134715.1 - 2 - 3 564 81
AF136293.1 - 2 - 3 420 81
AF186114.1 - 2 - 3 577 81
AK309629.1 - 3 - 4 457 81
AY129225.1 - 2 - 3 420 81
AY129226.1 - 2 - 3 189 81
AY129227.1 - 1 - 2 87 81
AY129228.1 - 1 - 2 39 81
AY358881.1 - 2 - 3 589 81
BC020674.1 - 2 - 3 599 81
CR541818.1 - 2 - 3 420 81
HM636064.1 - 2 - 3 420 81

View Full Product Details