Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SFSWAP
Entrez Gene ID
6433
Gene Name
splicing factor SWAP homolog
Gene Aliases
SFRS8, SWAP
Chromosome Location
Chr.12: 131711087 - 131799738 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001261411.1 NP_001248340.1 11 - 12 1859 81
NM_004592.3 NP_004583.2 11 - 12 1859 81
XM_011538654.2 XP_011536956.1 11 - 12 1811 81
XM_011538655.2 XP_011536957.1 11 - 12 1856 81
XM_017019799.1 XP_016875288.1 11 - 12 2298 81
GenBank mRNA AB208921.1 - 9 - 10 5050 81
BC008707.2 - 11 - 12 1856 81
BC015953.2 - 11 - 12 1856 81
BC024041.1 - 11 - 12 1857 81
BC136678.1 - 11 - 12 1809 81
BC144364.1 - 11 - 12 1809 81
U08377.1 - 11 - 12 1836 81

View Full Product Details