Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SFSWAP
Entrez Gene ID
6433
Gene Name
splicing factor SWAP homolog
Gene Aliases
SFRS8, SWAP
Chromosome Location
Chr.12: 131711087 - 131799738 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001261411.1 NP_001248340.1 1 - 2 356 78
NM_004592.3 NP_004583.2 1 - 2 356 78
XM_011538653.1 XP_011536955.1 1 - 2 353 78
XM_011538654.2 XP_011536956.1 1 - 2 355 78
XM_011538655.2 XP_011536957.1 1 - 2 353 78
XM_017019798.1 XP_016875287.1 1 - 2 353 78
XM_017019799.1 XP_016875288.1 1 - 2 328 78
GenBank mRNA BC008707.2 - 1 - 2 353 78
BC015953.2 - 1 - 2 353 78
BC024041.1 - 1 - 2 354 78
BC113420.1 - 1 - 2 306 78
BC136678.1 - 1 - 2 306 78
BC144364.1 - 1 - 2 306 78
U08377.1 - 1 - 2 333 78

View Full Product Details