The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000048.3 | NP_000039.2 | 11 - 12 | 943 | 104 |
NM_001024943.1 | NP_001020114.1 | 10 - 11 | 1067 | 104 | |
NM_001024944.1 | NP_001020115.1 | 10 - 11 | 1067 | 104 | |
NM_001024946.1 | NP_001020117.1 | 9 - 10 | 989 | 104 | |
GenBank mRNA | BC008195.1 | - | 11 - 12 | 905 | 104 |
BC033146.1 | - | 10 - 11 | 878 | 104 | |
J03058.1 | - | 11 - 12 | 901 | 104 | |
KU177920.1 | - | 9 - 10 | 832 | 104 | |
KU177921.1 | - | 8 - 9 | 754 | 104 | |
KU177922.1 | - | 5 - 6 | 517 | 104 | |
M14218.1 | - | 10 - 11 | 946 | 104 | |
M57638.1 | - | 10 - 11 | 877 | 104 | |
Y00753.1 | - | 10 - 11 | 877 | 104 |