The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000048.3 | NP_000039.2 | 12 - 13 | 1028 | 81 |
NM_001024943.1 | NP_001020114.1 | 11 - 12 | 1152 | 81 | |
NM_001024946.1 | NP_001020117.1 | 10 - 11 | 1074 | 81 | |
GenBank mRNA | BC008195.1 | - | 12 - 13 | 990 | 81 |
BC033146.1 | - | 11 - 12 | 963 | 81 | |
J03058.1 | - | 12 - 13 | 986 | 81 | |
KU177920.1 | - | 10 - 11 | 917 | 81 | |
KU177921.1 | - | 9 - 10 | 839 | 81 | |
KU177922.1 | - | 6 - 7 | 602 | 81 | |
M14218.1 | - | 11 - 12 | 1031 | 81 | |
M57638.1 | - | 11 - 12 | 962 | 81 | |
Y00753.1 | - | 11 - 12 | 962 | 81 |