The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000048.3 | NP_000039.2 | 13 - 14 | 1088 | 83 |
NM_001024943.1 | NP_001020114.1 | 12 - 13 | 1212 | 83 | |
NM_001024946.1 | NP_001020117.1 | 11 - 12 | 1134 | 83 | |
GenBank mRNA | BC008195.1 | - | 13 - 14 | 1050 | 83 |
BC033146.1 | - | 12 - 13 | 1023 | 83 | |
J03058.1 | - | 13 - 14 | 1046 | 83 | |
KU177920.1 | - | 11 - 12 | 977 | 83 | |
KU177921.1 | - | 10 - 11 | 899 | 83 | |
KU177922.1 | - | 7 - 8 | 662 | 83 | |
M14218.1 | - | 12 - 13 | 1091 | 83 | |
M57638.1 | - | 12 - 13 | 1022 | 83 | |
Y00753.1 | - | 12 - 13 | 1022 | 83 |