The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000048.3 | NP_000039.2 | 14 - 15 | 1175 | 55 |
NM_001024943.1 | NP_001020114.1 | 13 - 14 | 1299 | 55 | |
NM_001024944.1 | NP_001020115.1 | 12 - 13 | 1239 | 55 | |
NM_001024946.1 | NP_001020117.1 | 12 - 13 | 1221 | 55 | |
GenBank mRNA | BC008195.1 | - | 14 - 15 | 1137 | 55 |
BC033146.1 | - | 13 - 14 | 1110 | 55 | |
J03058.1 | - | 14 - 15 | 1133 | 55 | |
KU177920.1 | - | 12 - 13 | 1064 | 55 | |
KU177921.1 | - | 11 - 12 | 986 | 55 | |
KU177922.1 | - | 8 - 9 | 749 | 55 | |
M14218.1 | - | 13 - 14 | 1178 | 55 | |
M57638.1 | - | 13 - 14 | 1109 | 55 | |
Y00753.1 | - | 13 - 14 | 1109 | 55 |