The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000048.3 | NP_000039.2 | 7 - 8 | 628 | 122 |
NM_001024943.1 | NP_001020114.1 | 6 - 7 | 752 | 122 | |
NM_001024944.1 | NP_001020115.1 | 6 - 7 | 752 | 122 | |
GenBank mRNA | AK300519.1 | - | 7 - 8 | 583 | 122 |
AK316334.1 | - | 6 - 7 | 648 | 122 | |
BC008195.1 | - | 7 - 8 | 590 | 122 | |
BC033146.1 | - | 6 - 7 | 563 | 122 | |
BX328836.2 | - | 7 - 8 | 648 | 121 | |
J03058.1 | - | 7 - 8 | 586 | 122 | |
KU177920.1 | - | 5 - 6 | 517 | 122 | |
M14218.1 | - | 6 - 7 | 631 | 122 | |
M57638.1 | - | 6 - 7 | 562 | 122 | |
Y00753.1 | - | 6 - 7 | 562 | 122 |