The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000048.3 | NP_000039.2 | 8 - 9 | 718 | 104 |
NM_001024943.1 | NP_001020114.1 | 7 - 8 | 842 | 104 | |
NM_001024944.1 | NP_001020115.1 | 7 - 8 | 842 | 104 | |
GenBank mRNA | BC008195.1 | - | 8 - 9 | 680 | 104 |
BC033146.1 | - | 7 - 8 | 653 | 104 | |
BX328836.2 | - | 8 - 9 | 737 | 104 | |
J03058.1 | - | 8 - 9 | 676 | 104 | |
KU177920.1 | - | 6 - 7 | 607 | 104 | |
M14218.1 | - | 7 - 8 | 721 | 104 | |
M57638.1 | - | 7 - 8 | 652 | 104 | |
Y00753.1 | - | 7 - 8 | 652 | 104 |