Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RCC1
Entrez Gene ID
1104
Gene Name
regulator of chromosome condensation 1
Gene Aliases
CHC1, RCC1-I, SNHG3-RCC1
Chromosome Location
Chr.1: 28505943 - 28539196 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001048195.2 NP_001041660.1 3 - 4 210 71
GenBank mRNA BM546148.1 - 1 - 2 84 71
BQ436549.1 - 3 - 4 190 71
CV811053.1 - 3 - 4 181 71
KU177976.1 - 2 - 3 125 71

View Full Product Details