The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018963.4 | NP_061836.2 | 13 - 14 | 1539 | 144 |
NM_033656.3 | NP_387505.1 | 13 - 14 | 1539 | 144 | |
XM_011529611.1 | XP_011527913.1 | 13 - 14 | 1572 | 144 | |
XM_011529612.2 | XP_011527914.1 | 6 - 7 | 435 | 144 | |
XM_017028373.1 | XP_016883862.1 | 14 - 15 | 1344 | 144 | |
XM_017028375.1 | XP_016883864.1 | 13 - 14 | 1572 | 144 | |
GenBank mRNA | AB080586.1 | - | 13 - 14 | 1343 | 144 |
AB080587.1 | - | 13 - 14 | 1343 | 144 | |
AB080588.1 | - | 5 - 6 | 378 | 144 | |
AB080589.1 | - | 5 - 6 | 378 | 144 | |
AB080591.1 | - | 5 - 6 | 378 | 144 | |
AB080592.1 | - | 5 - 6 | 378 | 144 | |
AB080593.1 | - | 5 - 6 | 378 | 144 | |
AJ238214.2 | - | 13 - 14 | 1343 | 144 | |
AJ292465.1 | - | 13 - 14 | 1323 | 144 | |
AJ292466.1 | - | 13 - 14 | 1323 | 144 |