The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018963.4 | NP_061836.2 | 28 - 29 | 3612 | 141 |
NM_033656.3 | NP_387505.1 | 28 - 29 | 3612 | 141 | |
XM_011529611.1 | XP_011527913.1 | 28 - 29 | 3645 | 141 | |
XM_011529612.2 | XP_011527914.1 | 21 - 22 | 2508 | 141 | |
XM_011529613.1 | XP_011527915.1 | 6 - 7 | 607 | 141 | |
XM_017028373.1 | XP_016883862.1 | 29 - 30 | 3417 | 141 | |
XM_017028374.1 | XP_016883863.1 | 4 - 5 | 2247 | 141 | |
GenBank mRNA | AB080586.1 | - | 28 - 29 | 3416 | 141 |
AB080587.1 | - | 28 - 29 | 3416 | 141 | |
AB080590.1 | - | 3 - 4 | 273 | 141 | |
AJ238214.2 | - | 28 - 29 | 3416 | 141 | |
AJ292465.1 | - | 28 - 29 | 3396 | 141 | |
AJ292466.1 | - | 28 - 29 | 3396 | 141 |