The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018963.4 | NP_061836.2 | 31 - 32 | 3955 | 133 |
NM_033656.3 | NP_387505.1 | 31 - 32 | 3955 | 133 | |
XM_011529611.1 | XP_011527913.1 | 31 - 32 | 3988 | 133 | |
XM_011529612.2 | XP_011527914.1 | 24 - 25 | 2851 | 133 | |
XM_011529613.1 | XP_011527915.1 | 9 - 10 | 950 | 133 | |
XM_017028373.1 | XP_016883862.1 | 32 - 33 | 3760 | 133 | |
XM_017028374.1 | XP_016883863.1 | 7 - 8 | 2590 | 133 | |
GenBank mRNA | AB080586.1 | - | 31 - 32 | 3759 | 133 |
AB080587.1 | - | 31 - 32 | 3759 | 133 | |
AB080590.1 | - | 6 - 7 | 616 | 133 | |
AJ238214.2 | - | 31 - 32 | 3759 | 133 | |
AJ292465.1 | - | 31 - 32 | 3739 | 133 | |
AJ292466.1 | - | 31 - 32 | 3739 | 133 |