Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BRWD1
Entrez Gene ID
54014
Gene Name
bromodomain and WD repeat domain containing 1
Gene Aliases
C21orf107, DCAF19, N143, WDR9
Chromosome Location
Chr.21: 39184176 - 39321595 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018963.4 NP_061836.2 32 - 33 4078 152
NM_033656.3 NP_387505.1 32 - 33 4078 152
XM_011529611.1 XP_011527913.1 32 - 33 4111 152
XM_011529612.2 XP_011527914.1 25 - 26 2974 152
XM_011529613.1 XP_011527915.1 10 - 11 1073 152
XM_017028373.1 XP_016883862.1 33 - 34 3883 152
XM_017028374.1 XP_016883863.1 8 - 9 2713 152
GenBank mRNA AB080586.1 - 32 - 33 3882 152
AB080587.1 - 32 - 33 3882 152
AB080590.1 - 7 - 8 739 152
AJ238214.2 - 32 - 33 3882 152
AJ292465.1 - 32 - 33 3862 152
AJ292466.1 - 32 - 33 3862 152
AK125906.1 - 16 - 17 2030 152

View Full Product Details