The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018051.4 | NP_060521.4 | 15 - 16 | 2075 | 64 |
XM_005249549.1 | XP_005249606.1 | 12 - 13 | 1667 | 64 | |
XM_006716041.2 | XP_006716104.1 | 15 - 16 | 2099 | 64 | |
XM_011516367.1 | XP_011514669.1 | 15 - 16 | 1941 | 64 | |
XM_011516368.2 | XP_011514670.1 | 15 - 16 | 2099 | 64 | |
XM_011516369.2 | XP_011514671.1 | 15 - 16 | 1930 | 64 | |
XM_017012381.1 | XP_016867870.1 | 15 - 16 | 2121 | 64 | |
XM_017012382.1 | XP_016867871.1 | 15 - 16 | 2090 | 64 | |
XM_017012383.1 | XP_016867872.1 | 15 - 16 | 2151 | 64 | |
XM_017012384.1 | XP_016867873.1 | 12 - 13 | 1658 | 64 | |
XM_017012385.1 | XP_016867874.1 | 14 - 15 | 1832 | 64 | |
XM_017012386.1 | XP_016867875.1 | 12 - 13 | 1611 | 64 | |
XM_017012387.1 | XP_016867876.1 | 15 - 16 | 1951 | 64 | |
XM_017012388.1 | XP_016867877.1 | 5 - 6 | 835 | 64 | |
GenBank mRNA | AK027228.1 | - | 4 - 5 | 432 | 64 |
BC014491.1 | - | 15 - 16 | 2075 | 64 | |
CR627036.1 | - | 10 - 11 | 3717 | 64 |