The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005872.2 | NP_005863.1 | 1 - 2 | 149 | 89 |
GenBank mRNA | AB020623.1 | - | 1 - 2 | 144 | 89 |
AF081788.1 | - | 1 - 2 | 96 | 89 | |
AK222637.1 | - | 1 - 2 | 126 | 89 | |
AK313141.1 | - | 1 - 2 | 129 | 89 | |
BC005285.1 | - | 1 - 2 | 118 | 89 | |
BC012623.1 | - | 1 - 2 | 122 | 89 | |
BC022880.1 | - | 1 - 2 | 131 | 89 | |
BC039238.1 | - | 1 - 2 | 120 | 89 | |
BT019390.1 | - | 1 - 2 | 96 | 89 | |
CR542037.1 | - | 1 - 2 | 96 | 89 |