The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001321468.1 | NP_001308397.1 | 2 - 3 | 294 | 109 |
NM_001321474.1 | NP_001308403.1 | 2 - 3 | 294 | 109 | |
NM_001321478.1 | NP_001308407.1 | 2 - 3 | 294 | 109 | |
NM_001321481.1 | NP_001308410.1 | 2 - 3 | 294 | 109 | |
NM_025264.4 | NP_079540.2 | 2 - 3 | 294 | 109 | |
NR_028102.1 | 2 - 3 | 294 | 109 | ||
NR_135659.1 | 2 - 3 | 294 | 109 | ||
NR_135660.1 | 2 - 3 | 294 | 109 | ||
XM_017005050.1 | XP_016860539.1 | 2 - 3 | 294 | 109 | |
XM_017005051.1 | XP_016860540.1 | 2 - 3 | 294 | 109 | |
XM_017005052.1 | XP_016860541.1 | 2 - 3 | 294 | 109 | |
XM_017005053.1 | XP_016860542.1 | 2 - 3 | 294 | 109 | |
XM_017005054.1 | XP_016860543.1 | 2 - 3 | 285 | 109 | |
GenBank mRNA | AK093580.1 | - | 2 - 3 | 265 | 109 |
AK226034.1 | - | 2 - 3 | 245 | 109 | |
AK292002.1 | - | 2 - 3 | 279 | 109 | |
AK301353.1 | - | 2 - 3 | 170 | 109 | |
AK307754.1 | - | 2 - 3 | 258 | 109 | |
AL832115.1 | - | 2 - 3 | 261 | 109 | |
BC004163.2 | - | 2 - 3 | 255 | 109 | |
BC013299.2 | - | 2 - 3 | 255 | 109 | |
DC379776.1 | - | 2 - 3 | 296 | 109 |