Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
THUMPD2
Entrez Gene ID
80745
Gene Name
THUMP domain containing 2
Gene Aliases
C2orf8
Chromosome Location
Chr.2: 39736060 - 39779276 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001321468.1 NP_001308397.1 5 - 6 838 125
NM_001321469.1 NP_001308398.1 5 - 6 828 125
NM_001321470.1 NP_001308399.1 5 - 6 780 125
NM_001321474.1 NP_001308403.1 5 - 6 838 125
NM_001321475.1 NP_001308404.1 5 - 6 828 125
NM_001321477.1 NP_001308406.1 6 - 7 1021 125
NM_001321478.1 NP_001308407.1 6 - 7 1129 125
NM_001321479.1 NP_001308408.1 4 - 5 428 125
NM_001321480.1 NP_001308409.1 6 - 7 1119 125
NM_001321481.1 NP_001308410.1 6 - 7 1031 125
NM_025264.4 NP_079540.2 5 - 6 838 125
NR_028102.1 5 - 6 838 125
NR_135659.1 6 - 7 1129 125
NR_135660.1 6 - 7 1031 125
NR_135661.1 6 - 7 1119 125
NR_135662.1 5 - 6 828 125
NR_135663.1 4 - 5 428 125
NR_135664.1 6 - 7 1021 125
XM_017005050.1 XP_016860539.1 5 - 6 790 125
XM_017005051.1 XP_016860540.1 5 - 6 790 125
XM_017005054.1 XP_016860543.1 5 - 6 1519 125
GenBank mRNA AF380577.1 - 2 - 3 134 125
AF380578.1 - 2 - 3 1642 125
AF380579.1 - 2 - 3 808 125
AK057801.1 - 3 - 4 220 125
AK093580.1 - 5 - 6 809 125
AK226034.1 - 5 - 6 790 125
AK292002.1 - 5 - 6 823 125
AK298178.1 - 5 - 6 725 125
AK301353.1 - 6 - 7 907 125
AL832115.1 - 5 - 6 805 125
BC004163.2 - 5 - 6 799 125
BC013299.2 - 5 - 6 799 125

View Full Product Details