The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001321469.1 | NP_001308398.1 | 8 - 9 | 1112 | 171 |
NM_001321470.1 | NP_001308399.1 | 8 - 9 | 1064 | 171 | |
NM_001321477.1 | NP_001308406.1 | 9 - 10 | 1305 | 171 | |
NM_001321478.1 | NP_001308407.1 | 9 - 10 | 1413 | 171 | |
NM_001321479.1 | NP_001308408.1 | 7 - 8 | 712 | 171 | |
NM_001321480.1 | NP_001308409.1 | 9 - 10 | 1403 | 171 | |
NM_001321481.1 | NP_001308410.1 | 9 - 10 | 1315 | 171 | |
NM_025264.4 | NP_079540.2 | 8 - 9 | 1122 | 171 | |
NR_028102.1 | 9 - 10 | 1243 | 171 | ||
NR_135659.1 | 10 - 11 | 1534 | 171 | ||
NR_135660.1 | 10 - 11 | 1436 | 171 | ||
NR_135661.1 | 10 - 11 | 1524 | 171 | ||
NR_135662.1 | 9 - 10 | 1233 | 171 | ||
NR_135663.1 | 8 - 9 | 833 | 171 | ||
NR_135664.1 | 10 - 11 | 1426 | 171 | ||
XM_011533119.2 | XP_011531421.1 | 5 - 6 | 477 | 171 | |
XM_017005050.1 | XP_016860539.1 | 8 - 9 | 1074 | 171 | |
XM_017005054.1 | XP_016860543.1 | 8 - 9 | 1803 | 171 | |
GenBank mRNA | AF380577.1 | - | 6 - 7 | 539 | 171 |
AF380578.1 | - | 6 - 7 | 2047 | 171 | |
AF380579.1 | - | 6 - 7 | 1213 | 171 | |
AK057801.1 | - | 7 - 8 | 625 | 171 | |
AK093580.1 | - | 9 - 10 | 1214 | 171 | |
AK226034.1 | - | 8 - 9 | 1074 | 171 | |
AK292002.1 | - | 9 - 10 | 1228 | 171 | |
AK298178.1 | - | 8 - 9 | 1009 | 171 | |
AL832115.1 | - | 9 - 10 | 1210 | 171 | |
BC004163.2 | - | 8 - 9 | 1083 | 171 | |
BC013299.2 | - | 8 - 9 | 1083 | 171 |